Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001085445.3 | XP_001085445.1 | 656 | 67 | ||
XM_015114062.2 | XP_014969548.1 | 901 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |