Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MICU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001089388.4 | XP_001089388.3 | 531 | 121 | ||
XM_015120728.2 | XP_014976214.2 | 516 | 121 | |||
XM_028837518.1 | XP_028693351.1 | 569 | 121 | |||
GenBank mRNA | JU333393.1 | - | 461 | 121 | ||
JV046451.1 | - | 473 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |