Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266682.1 | NP_001253611.1 | 797 | 77 | ||
XM_015146191.2 | XP_015001677.1 | 889 | 77 | |||
XM_028852165.1 | XP_028707998.1 | 1110 | 77 | |||
GenBank mRNA | JU336679.1 | - | 776 | 77 | ||
JU336680.1 | - | 772 | 77 | |||
JU477476.1 | - | 849 | 77 | |||
JU477477.1 | - | 845 | 77 | |||
JV048427.1 | - | 801 | 77 | |||
JV048428.1 | - | 797 | 77 | |||
JV630765.1 | - | 871 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |