Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABGGTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261560.2 | NP_001248489.1 | 141 | 118 | ||
GenBank mRNA | JU335736.1 | - | 168 | 118 | ||
JU473413.1 | - | 141 | 118 | |||
JV490939.1 | - | 172 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |