Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YPEL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194271.3 | NP_001181200.1 | 525 | 67 | ||
XM_015112018.2 | XP_014967504.1 | 524 | 67 | |||
XM_015112019.2 | XP_014967505.1 | 380 | 67 | |||
XM_028831477.1 | XP_028687310.1 | 716 | 67 | |||
GenBank mRNA | JU319726.1 | - | 280 | 67 | ||
JU319727.1 | - | 289 | 67 | |||
JU473405.1 | - | 302 | 67 | |||
JU473406.1 | - | 494 | 67 | |||
JV043675.1 | - | 281 | 67 | |||
JV566026.1 | - | 342 | 67 | |||
JV619091.1 | - | 306 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |