Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPY30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261093.1 | NP_001248022.1 | 79 | 145 | ||
XM_015112025.2 | XP_014967511.1 | 192 | 145 | |||
XM_015112026.2 | XP_014967512.1 | 192 | 145 | |||
XM_015112027.2 | XP_014967513.1 | 199 | 145 | |||
XM_015112028.2 | XP_014967514.1 | 195 | 145 | |||
XM_015112029.2 | XP_014967515.1 | 246 | 145 | |||
XM_015112030.2 | XP_014967516.1 | 308 | 145 | |||
XM_028831479.1 | XP_028687312.1 | 192 | 145 | |||
XM_028831481.1 | XP_028687314.1 | 211 | 145 | |||
XM_028831482.1 | XP_028687315.1 | 192 | 145 | |||
XM_028831484.1 | XP_028687317.1 | 191 | 145 | |||
GenBank mRNA | JU320223.1 | - | 238 | 145 | ||
JU473496.1 | - | 76 | 145 | |||
JV048468.1 | - | 79 | 145 | |||
JV639282.1 | - | 250 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |