Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266533.2 | NP_001253462.1 | 146 | 55 | ||
XM_015131546.2 | XP_014987032.1 | 237 | 55 | |||
GenBank mRNA | JU325229.1 | - | 78 | 55 | ||
JU473772.1 | - | 131 | 55 | |||
JV636173.1 | - | 148 | 55 | |||
JV715754.1 | - | 100 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |