Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194599.2 | NP_001181528.1 | 534 | 72 | ||
XM_028843394.1 | XP_028699227.1 | 467 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |