Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001113439.4 | XP_001113439.2 | 693 | 101 | ||
XM_028839007.1 | XP_028694840.1 | 693 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |