Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAB39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267016.1 | NP_001253945.1 | 515 | 125 | ||
XM_015111248.2 | XP_014966734.1 | 574 | 125 | |||
GenBank mRNA | JU319865.1 | - | 512 | 125 | ||
JU475973.1 | - | 491 | 125 | |||
JV045169.1 | - | 515 | 125 | |||
JV601991.1 | - | 715 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |