Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194638.2 | NP_001181567.1 | 334 | 65 | ||
XM_015113721.2 | XP_014969207.1 | 437 | 65 | |||
GenBank mRNA | JV602491.1 | - | 363 | 65 | ||
JV635720.1 | - | 312 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |