Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001086242.4 | XP_001086242.2 | 525 | 89 | ||
XM_015114693.2 | XP_014970179.2 | 547 | 89 | |||
XM_028833578.1 | XP_028689411.1 | 630 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |