Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015126450.2 | XP_014981936.1 | 731 | 77 | ||
XM_015126451.2 | XP_014981937.1 | 579 | 77 | |||
XM_028841542.1 | XP_028697375.1 | 504 | 77 | |||
XM_028841543.1 | XP_028697376.1 | 374 | 77 | |||
XM_028841544.1 | XP_028697377.1 | 388 | 77 | |||
GenBank mRNA | JV064396.1 | - | 322 | 77 | ||
JV472713.1 | - | 314 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |