Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193916.2 | NP_001180845.1 | 157 | 114 | ||
XM_001091708.4 | XP_001091708.1 | 410 | 114 | |||
XM_028828817.1 | XP_028684650.1 | 221 | 114 | |||
XM_028828818.1 | XP_028684651.1 | 383 | 114 | |||
GenBank mRNA | JU320856.1 | - | 380 | 114 | ||
JU472984.1 | - | 329 | 114 | |||
JV044318.1 | - | 268 | 114 | |||
JV490012.1 | - | 381 | 114 | |||
JV529797.1 | - | 299 | 114 | |||
JV602575.1 | - | 327 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |