Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266356.1 | NP_001253285.1 | 661 | 107 | ||
XM_015140114.2 | XP_014995600.1 | 817 | 107 | |||
XM_015140117.2 | XP_014995603.1 | 544 | 107 | |||
XM_015140118.2 | XP_014995604.1 | 585 | 107 | |||
GenBank mRNA | JU336462.1 | - | 813 | 107 | ||
JU472540.1 | - | 792 | 107 | |||
JV044090.1 | - | 661 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |