Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INHBE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001115958.3 | XP_001115958.2 | 1410 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta Inventoried |