Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194016.2 | NP_001180945.2 | 151 | 177 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |