Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266807.1 | NP_001253736.1 | 2110 | 144 | ||
XM_015146991.2 | XP_015002477.1 | 619 | 144 | |||
GenBank mRNA | JU323415.1 | - | 586 | 144 | ||
JU476379.1 | - | 2110 | 144 | |||
JV047624.1 | - | 2110 | 144 | |||
JV637781.1 | - | 609 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |