Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015137465.2 | XP_014992951.2 | 446 | 110 | ||
XM_015137466.2 | XP_014992952.2 | 255 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |