Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARVELD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001094419.4 | XP_001094419.1 | 1609 | 149 | ||
XM_015140247.2 | XP_014995733.1 | 2027 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |