Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYW5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193883.2 | NP_001180812.2 | 342 | 82 | ||
GenBank mRNA | JU338818.1 | - | 322 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |