Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032807.2 | NP_001027979.1 | 178 | 89 | ||
XM_028846608.1 | XP_028702441.1 | 347 | 89 | |||
XM_028846609.1 | XP_028702442.1 | 347 | 89 | |||
XM_028846610.1 | XP_028702443.1 | 347 | 89 | |||
GenBank mRNA | AF016372.1 | - | 181 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |