Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCHR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032948.1 | NP_001028120.1 | 587 | 158 | ||
XM_015136945.2 | XP_014992431.1 | 1049 | 158 | |||
XM_015136946.1 | XP_014992432.1 | 888 | 158 | |||
XM_015136947.2 | XP_014992433.1 | 712 | 158 | |||
GenBank mRNA | AF513989.1 | - | 587 | 158 | ||
AY078246.1 | - | 578 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Macaca mulatta |