Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHI3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265920.1 | NP_001252849.1 | 1142 | 90 | ||
GenBank mRNA | JU323002.1 | - | 1142 | 90 | ||
JV043861.1 | - | 1156 | 90 | |||
JV704305.1 | - | 1163 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |