Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015136681.2 | XP_014992167.1 | 1949 | 148 | ||
XM_015136688.2 | XP_014992174.1 | 1914 | 148 | |||
XM_028835304.1 | XP_028691137.1 | 1535 | 148 | |||
GenBank mRNA | JU322260.1 | - | 1483 | 148 | ||
JU474436.1 | - | 1505 | 148 | |||
JV046270.1 | - | 1534 | 148 | |||
JV643321.1 | - | 1306 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |