Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHHADH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257492.1 | NP_001244421.1 | 218 | 70 | ||
GenBank mRNA | JU323220.1 | - | 218 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |