Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266518.1 | NP_001253447.1 | 1188 | 92 | ||
XM_015114984.2 | XP_014970470.1 | 3370 | 92 | |||
GenBank mRNA | JU320953.1 | - | 1188 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |