Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194910.2 | NP_001181839.1 | 506 | 77 | ||
XM_001102167.4 | XP_001102167.1 | 504 | 77 | |||
XM_015126960.2 | XP_014982446.1 | 504 | 77 | |||
XM_015126961.2 | XP_014982447.1 | 504 | 77 | |||
XM_015126962.2 | XP_014982448.1 | 504 | 77 | |||
XM_028841826.1 | XP_028697659.1 | 285 | 77 | |||
GenBank mRNA | JU320549.1 | - | 225 | 77 | ||
JU336951.1 | - | 225 | 77 | |||
JU470839.1 | - | 178 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |