Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARSF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015126895.2 | XP_014982381.2 | 1434 | 128 | ||
XM_028842198.1 | XP_028698031.1 | 1348 | 128 | |||
XM_028842199.1 | XP_028698032.1 | 1465 | 128 | |||
XM_028842200.1 | XP_028698033.1 | 1459 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |