Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194693.2 | NP_001181622.1 | 1447 | 129 | ||
XM_001115852.4 | XP_001115852.1 | 1477 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |