Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTP4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193694.2 | NP_001180623.1 | 273 | 117 | ||
XM_015146081.2 | XP_015001567.1 | 1175 | 117 | |||
XM_015146083.2 | XP_015001569.1 | 1138 | 117 | |||
XM_015146084.2 | XP_015001570.1 | 1145 | 117 | |||
XM_015146085.2 | XP_015001571.1 | 1175 | 117 | |||
XM_028852114.1 | XP_028707947.1 | 1295 | 117 | |||
XM_028852115.1 | XP_028707948.1 | 1252 | 117 | |||
GenBank mRNA | JU470942.1 | - | 431 | 117 | ||
JV046058.1 | - | 537 | 117 | |||
JV635829.1 | - | 529 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |