Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266811.1 | NP_001253740.1 | 522 | 141 | ||
XM_015145694.2 | XP_015001180.1 | 563 | 141 | |||
GenBank mRNA | JU319935.1 | - | 522 | 141 | ||
JV566130.1 | - | 566 | 141 | |||
JV601984.1 | - | 578 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |