Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYND19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194731.2 | NP_001181660.1 | 344 | 82 | ||
GenBank mRNA | JU332828.1 | - | 153 | 82 | ||
JU474239.1 | - | 272 | 82 | |||
JV046132.1 | - | 144 | 82 | |||
JV601388.1 | - | 191 | 82 | |||
JV638766.1 | - | 307 | 82 | |||
JV672360.1 | - | 162 | 82 |