Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258135.2 | NP_001245064.1 | 995 | 115 | ||
GenBank mRNA | JU324394.1 | - | 954 | 115 | ||
JU473777.1 | - | 997 | 115 | |||
JV045395.1 | - | 978 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |