Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082965.1 | NP_001076434.1 | 282 | 105 | ||
GenBank mRNA | EF426304.1 | - | 282 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |