Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACOT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266088.1 | NP_001253017.1 | 220 | 82 | ||
XM_015148852.2 | XP_015004338.1 | 243 | 82 | |||
GenBank mRNA | JU327233.1 | - | 2030 | 82 | ||
JU472253.1 | - | 220 | 82 | |||
JV597667.1 | - | 2544 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |