Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G3BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261742.1 | NP_001248671.1 | 1010 | 151 | ||
XM_015141262.2 | XP_014996748.1 | 992 | 151 | |||
XM_015141264.2 | XP_014996750.1 | 920 | 151 | |||
XM_028849271.1 | XP_028705104.1 | 1267 | 151 | |||
GenBank mRNA | JU471056.1 | - | 979 | 151 | ||
JU471057.1 | - | 1010 | 151 | |||
JV637223.1 | - | 1010 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |