Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAMU-DMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193474.1 | NP_001180403.1 | 426 | 129 | ||
GenBank mRNA | CO725887.1 | - | 429 | 129 | ||
CO726452.1 | - | 382 | 129 | |||
JU325668.1 | - | 121 | 129 | |||
JV049020.1 | - | 418 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta Inventoried |