Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BATF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266082.1 | NP_001253011.1 | 285 | 82 | ||
GenBank mRNA | JU473620.1 | - | 285 | 82 | ||
JV569069.1 | - | 287 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |