Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST3GAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266341.1 | NP_001253270.1 | 1792 | 135 | ||
XM_028840536.1 | XP_028696369.1 | 1893 | 135 | |||
GenBank mRNA | JU332128.1 | - | 1662 | 135 | ||
JU470852.1 | - | 1801 | 135 | |||
JV044250.1 | - | 1792 | 135 | |||
JV567663.1 | - | 1849 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |