Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194502.3 | NP_001181431.2 | 223 | 127 | ||
GenBank mRNA | JU327025.1 | - | 200 | 127 | ||
JV049568.1 | - | 204 | 127 | |||
JV565029.1 | - | 211 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |