Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUCO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001100732.4 | XP_001100732.2 | 1301 | 105 | ||
XM_015124042.2 | XP_014979528.2 | 1043 | 105 | |||
XM_028850415.1 | XP_028706248.1 | 1307 | 105 | |||
GenBank mRNA | JU334889.1 | - | 1252 | 105 | ||
JU334890.1 | - | 1252 | 105 | |||
JU334891.1 | - | 1252 | 105 | |||
JU475899.1 | - | 1199 | 105 | |||
JV047300.1 | - | 1258 | 105 | |||
JV047301.1 | - | 1258 | 105 | |||
JV636208.1 | - | 1215 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |