Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM20B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260759.1 | NP_001247688.1 | 986 | 74 | ||
XM_015121554.2 | XP_014977040.1 | 3017 | 74 | |||
XM_015121565.2 | XP_014977051.1 | 1934 | 74 | |||
XM_028845765.1 | XP_028701598.1 | 890 | 74 | |||
GenBank mRNA | JU337385.1 | - | 930 | 74 | ||
JU473000.1 | - | 986 | 74 | |||
JV045248.1 | - | 986 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |