Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JOSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266873.1 | NP_001253802.1 | 673 | 93 | ||
XM_015150318.2 | XP_015005804.1 | 1258 | 93 | |||
XM_015150323.2 | XP_015005809.1 | 742 | 93 | |||
GenBank mRNA | JU320086.1 | - | 769 | 93 | ||
JU332160.1 | - | 769 | 93 | |||
JU332161.1 | - | 769 | 93 | |||
JU478086.1 | - | 674 | 93 | |||
JV048862.1 | - | 673 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |