Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NELFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015116253.2 | XP_014971739.1 | 2289 | 100 | ||
GenBank mRNA | JU333872.1 | - | 643 | 100 | ||
JU333873.1 | - | 643 | 100 | |||
JU472033.1 | - | 664 | 100 | |||
JU472034.1 | - | 664 | 100 | |||
JV046684.1 | - | 794 | 100 | |||
JV046685.1 | - | 794 | 100 |