Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260989.1 | NP_001247918.1 | 269 | 65 | ||
XM_015141777.2 | XP_014997263.1 | 325 | 65 | |||
GenBank mRNA | JU334832.1 | - | 205 | 65 | ||
JU475859.1 | - | 269 | 65 | |||
JV716396.1 | - | 272 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |