Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261280.1 | NP_001248209.1 | 1034 | 98 | ||
XM_015138149.2 | XP_014993635.1 | 5168 | 98 | |||
XM_015138150.2 | XP_014993636.1 | 919 | 98 | |||
XM_015138151.2 | XP_014993637.1 | 957 | 98 | |||
GenBank mRNA | JU334842.1 | - | 947 | 98 | ||
JU472395.1 | - | 1047 | 98 | |||
JV047272.1 | - | 1034 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |