Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194726.3 | NP_001181655.3 | 849 | 125 | ||
GenBank mRNA | JU322648.1 | - | 873 | 125 | ||
JU476294.1 | - | 863 | 125 | |||
JV638384.1 | - | 882 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |