Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC4H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261736.1 | NP_001248665.1 | 108 | 101 | ||
XM_015127582.2 | XP_014983068.1 | 176 | 101 | |||
GenBank mRNA | JU335463.1 | - | 163 | 101 | ||
JU476405.1 | - | 108 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |