Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194335.1 | NP_001181264.1 | 198 | 130 | ||
GenBank mRNA | JU323383.1 | - | 194 | 130 | ||
JU476410.1 | - | 196 | 130 | |||
JV045474.1 | - | 179 | 130 | |||
JV638009.1 | - | 195 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |